Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARRDC3
Entrez Gene ID
541123
Gene Name
arrestin domain containing 3
Gene Aliases
-
Chromosome Location
Chr.7: 90839580 - 90853625 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076257.2 NP_001069725.1 5 - 6 1102 113
GenBank mRNA BC120276.1 - 5 - 6 1103 113
KP292570.1 - 5 - 6 1103 113

View Full Product Details