Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC8A
Entrez Gene ID
505605
Gene Name
leucine rich repeat containing 8 VRAC subunit A
Gene Aliases
LLRC8A
Chromosome Location
Chr.11: 99393986 - 99421580 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076807.1 NP_001070275.1 3 - 4 2364 89
GenBank mRNA BC123432.1 - 3 - 4 2364 89

View Full Product Details