Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADGRL4
Entrez Gene ID
535066
Gene Name
adhesion G protein-coupled receptor L4
Gene Aliases
ELTD1
Chromosome Location
Chr.3: 65667452 - 65808369 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076908.2 NP_001070376.1 10 - 11 1542 124
GenBank mRNA BC123530.1 - 10 - 11 1543 124

View Full Product Details