Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACOT8
Entrez Gene ID
504360
Gene Name
acyl-CoA thioesterase 8
Gene Aliases
-
Chromosome Location
Chr.13: 74608960 - 74620968 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075157.2 NP_001068625.2 5 - 6 927 134

View Full Product Details