Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OOEP
Entrez Gene ID
507401
Gene Name
oocyte expressed protein
Gene Aliases
-
Chromosome Location
Chr.9: 12765347 - 12766525 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077869.2 NP_001071337.1 2 - 3 429 107
GenBank mRNA BC126853.1 - 2 - 3 430 107

View Full Product Details