Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LY6G6E
Entrez Gene ID
512743
Gene Name
lymphocyte antigen 6 complex, locus G6E
Gene Aliases
-
Chromosome Location
Chr.23: 27596707 - 27598654 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081520.2 NP_001074989.1 3 - 4 298 75
GenBank mRNA BC133429.1 - 3 - 4 300 75
KU253661.1 - 2 - 3 179 75

View Full Product Details