Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAPZA1
Entrez Gene ID
534656
Gene Name
capping actin protein of muscle Z-line subunit alpha 1
Gene Aliases
-
Chromosome Location
Chr.3: 30657726 - 30701981 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083480.2 NP_001076949.1 2 - 3 143 118
GenBank mRNA BC114648.1 - 2 - 3 147 118
BT030727.1 - 2 - 3 104 118

View Full Product Details