Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL21A
Entrez Gene ID
615773
Gene Name
methyltransferase like 21A
Gene Aliases
FAM119A
Chromosome Location
Chr.2: 95908907 - 95918927 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083518.2 NP_001076987.1 2 - 3 296 65
GenBank mRNA BC123706.1 - 2 - 3 297 65

View Full Product Details