Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PGLYRP1
Entrez Gene ID
282305
Gene Name
peptidoglycan recognition protein 1
Gene Aliases
PGLYRP
Chromosome Location
Chr.18: 53513719 - 53515780 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174573.3 NP_776998.1 2 - 3 415 106
GenBank mRNA AY083309.1 - 2 - 3 417 106
BC142042.1 - 2 - 3 504 106

View Full Product Details