Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD38
Entrez Gene ID
327677
Gene Name
CD38 molecule
Gene Aliases
-
Chromosome Location
Chr.6: 110986420 - 111044390 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175798.3 NP_786992.1 3 - 4 599 103
GenBank mRNA AF101437.1 - 3 - 4 548 103
BC126708.1 - 3 - 4 599 103

View Full Product Details