Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM50A
Entrez Gene ID
515539
Gene Name
family with sequence similarity 50 member A
Gene Aliases
-
Chromosome Location
Chr.X: 37504050 - 37509805 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034496.2 NP_001029668.2 11 - 12 982 53
GenBank mRNA BC103028.1 - 10 - 11 1259 53
CO887857.1 - 3 - 4 202 53

View Full Product Details