Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBATA
Entrez Gene ID
515913
Gene Name
thymus, brain and testes associated
Gene Aliases
C28H10orf27, SPATIAL
Chromosome Location
Chr.28: 26937387 - 26952734 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038121.2 NP_001033210.2 7 - 8 974 102
GenBank mRNA BC110031.1 - 7 - 8 975 102
CK774174.1 - 2 - 3 272 102

View Full Product Details