Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MIS18A
Entrez Gene ID
510809
Gene Name
MIS18 kinetochore protein A
Gene Aliases
-
Chromosome Location
Chr.1: 3210652 - 3222939 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098010.2 NP_001091479.1 2 - 3 428 115
GenBank mRNA BC140629.1 - 2 - 3 429 115

View Full Product Details