Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRS3
Entrez Gene ID
509838
Gene Name
fibroblast growth factor receptor substrate 3
Gene Aliases
-
Chromosome Location
Chr.23: 15583410 - 15593147 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098918.2 NP_001092388.1 1 - 2 47 81
GenBank mRNA BC146158.1 - 1 - 2 48 81

View Full Product Details