Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDK20
Entrez Gene ID
510920
Gene Name
cyclin dependent kinase 20
Gene Aliases
CCRK
Chromosome Location
Chr.8: 90058696 - 90067693 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098933.1 NP_001092403.1 5 - 6 828 96
GenBank mRNA BC146216.1 - 5 - 6 828 96

View Full Product Details