Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JTB
Entrez Gene ID
513970
Gene Name
jumping translocation breakpoint
Gene Aliases
-
Chromosome Location
Chr.3: 16461551 - 16464005 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100360.1 NP_001093830.1 3 - 4 646 89
GenBank mRNA BC150014.1 - 3 - 4 646 89

View Full Product Details