Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALDOA
Entrez Gene ID
509566
Gene Name
aldolase, fructose-bisphosphate A
Gene Aliases
-
Chromosome Location
Chr.25: 26212564 - 26225472 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101915.1 NP_001095385.1 7 - 8 1081 133
GenBank mRNA BC148010.1 - 7 - 8 1081 133

View Full Product Details