Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A10
Entrez Gene ID
509728
Gene Name
solute carrier family 22, member 10
Gene Aliases
-
Chromosome Location
Chr.29: 41582419 - 41608529 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101917.1 NP_001095387.1 7 - 8 1292 78
GenBank mRNA BC151516.1 - 7 - 8 1292 78

View Full Product Details