Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
METRN
Entrez Gene ID
515528
Gene Name
meteorin, glial cell differentiation regulator
Gene Aliases
-
Chromosome Location
Chr.25: 614323 - 616436 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102026.2 NP_001095496.1 1 - 2 116 120
GenBank mRNA BC149635.1 - 1 - 2 117 120

View Full Product Details