Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TDP2
Entrez Gene ID
507579
Gene Name
tyrosyl-DNA phosphodiesterase 2
Gene Aliases
TTRAP
Chromosome Location
Chr.23: 33082523 - 33094886 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105341.1 NP_001098811.1 2 - 3 298 119
GenBank mRNA BC134603.1 - 2 - 3 298 119

View Full Product Details