Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPL52
Entrez Gene ID
509469
Gene Name
mitochondrial ribosomal protein L52
Gene Aliases
-
Chromosome Location
Chr.10: 21979455 - 21984650 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110180.2 NP_001103650.1 1 - 2 33 57
GenBank mRNA BC102378.1 - 1 - 2 36 57

View Full Product Details