Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM89
Entrez Gene ID
505670
Gene Name
transmembrane protein 89
Gene Aliases
-
Chromosome Location
Chr.22: 51262152 - 51263037 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111068.2 NP_001104538.1 1 - 2 310 79
GenBank mRNA BC142360.1 - 1 - 2 311 79

View Full Product Details