Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACBD7
Entrez Gene ID
508284
Gene Name
acyl-CoA binding domain containing 7
Gene Aliases
-
Chromosome Location
Chr.13: 29654008 - 29660465 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113297.3 NP_001106768.1 3 - 4 271 115
GenBank mRNA BC102899.1 - 3 - 4 242 115

View Full Product Details