Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS35L
Entrez Gene ID
534058
Gene Name
VPS35 endosomal protein sorting factor like
Gene Aliases
C16orf62, C25H16orf62
Chromosome Location
Chr.25: 17159011 - 17297111 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046368.2 NP_001039833.2 6 - 7 523 80
GenBank mRNA BC105446.1 - 6 - 7 559 80
DV883546.1 - 3 - 4 205 80

View Full Product Details