Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYT4
Entrez Gene ID
539867
Gene Name
synaptotagmin 4
Gene Aliases
-
Chromosome Location
Chr.24: 12650269 - 12660067 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098108.1 NP_001091577.1 3 - 4 1186 66
GenBank mRNA BC140533.1 - 3 - 4 1186 66

View Full Product Details