Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRAS
Entrez Gene ID
540803
Gene Name
muscle RAS oncogene homolog
Gene Aliases
-
Chromosome Location
Chr.1: 130707101 - 130774015 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098118.1 NP_001091587.1 2 - 3 285 99
GenBank mRNA BC140603.1 - 2 - 3 285 99
BC142259.1 - 2 - 3 382 99
BC142462.1 - 2 - 3 239 99

View Full Product Details