Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP2K4
Entrez Gene ID
526469
Gene Name
mitogen-activated protein kinase kinase 4
Gene Aliases
-
Chromosome Location
Chr.19: 30649010 - 30726941 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099038.1 NP_001092508.1 7 - 8 786 122
GenBank mRNA BC142195.1 - 7 - 8 786 122

View Full Product Details