Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NID1
Entrez Gene ID
534319
Gene Name
nidogen 1
Gene Aliases
-
Chromosome Location
Chr.28: 8687268 - 8784219 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101155.1 NP_001094625.1 5 - 6 1384 58
GenBank mRNA BC148974.1 - 5 - 6 1384 58

View Full Product Details