Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ANKRD9
Entrez Gene ID
532402
Gene Name
ankyrin repeat domain 9
Gene Aliases
-
Chromosome Location
Chr.21: 67248222 - 67254884 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102144.2 NP_001095614.1 1 - 2 181 151
GenBank mRNA BC151561.1 - 1 - 2 182 151

View Full Product Details