Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPDR1
Entrez Gene ID
615376
Gene Name
ependymin related 1
Gene Aliases
-
Chromosome Location
Chr.4: 49750583 - 49787424 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102288.2 NP_001095758.1 1 - 2 561 80
GenBank mRNA BC147972.1 - 1 - 2 562 80

View Full Product Details