Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYRM2
Entrez Gene ID
615821
Gene Name
LYR motif containing 2
Gene Aliases
-
Chromosome Location
Chr.9: 60553638 - 60555717 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113310.1 NP_001106781.1 2 - 3 234 90
GenBank mRNA BC109510.1 - 2 - 3 234 90

View Full Product Details