Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNT1
Entrez Gene ID
540662
Gene Name
Wnt family member 1
Gene Aliases
-
Chromosome Location
Chr.5: 30832513 - 30835596 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114191.1 NP_001107663.1 3 - 4 659 85

View Full Product Details