Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAMD14
Entrez Gene ID
785513
Gene Name
sterile alpha motif domain containing 14
Gene Aliases
-
Chromosome Location
Chr.19: 36526568 - 36544631 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105018.1 NP_001098488.1 6 - 7 1222 110
GenBank mRNA BC146255.1 - 6 - 7 1222 110

View Full Product Details