Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LHX1
Entrez Gene ID
786041
Gene Name
LIM homeobox 1
Gene Aliases
-
Chromosome Location
Chr.19: 13300723 - 13306530 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105447.1 NP_001098917.1 4 - 5 1666 97
GenBank mRNA BC153212.1 - 4 - 5 1666 97
BC154403.1 - 2 - 3 192 97

View Full Product Details