Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRTAM
Entrez Gene ID
781864
Gene Name
cytotoxic and regulatory T cell molecule
Gene Aliases
-
Chromosome Location
Chr.15: 33457290 - 33486131 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105457.1 NP_001098927.1 2 - 3 231 76
GenBank mRNA BC134770.1 - 2 - 3 231 76

View Full Product Details