Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RER1
Entrez Gene ID
100125922
Gene Name
retention in endoplasmic reticulum sorting receptor 1
Gene Aliases
-
Chromosome Location
Chr.16: 50442762 - 50452801 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105492.1 NP_001098962.1 6 - 7 532 72
GenBank mRNA BC141981.1 - 6 - 7 660 72
BC151447.1 - 6 - 7 532 72

View Full Product Details