Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
BLOC1S3
Entrez Gene ID
538539
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 3
Gene Aliases
-
Chromosome Location
Chr.18: 52826563 - 52833958 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099086.1 NP_001092556.1 - 877 149
GenBank mRNA BC142188.1 - - 877 149

View Full Product Details