Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BLOC1S4
Entrez Gene ID
518544
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 4
Gene Aliases
CNO
Chromosome Location
Chr.6: 113216655 - 113218144 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080731.2 NP_001074200.1 1 - 1 1152 148
GenBank mRNA BC120238.1 - 1 - 1 1154 148

View Full Product Details