Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTCFL
Entrez Gene ID
100144618
Gene Name
CCCTC-binding factor like
Gene Aliases
BORIS
Chromosome Location
Chr.13: 58657726 - 58682600 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123365.1 NP_001116837.1 8 - 9 1419 63
GenBank mRNA EU527855.1 - 8 - 9 1419 63

View Full Product Details