Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXW4
Entrez Gene ID
513977
Gene Name
F-box and WD repeat domain containing 4
Gene Aliases
-
Chromosome Location
Chr.26: 22357362 - 22440702 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101985.1 NP_001095455.1 7 - 8 1037 71
GenBank mRNA BC149096.1 - 7 - 8 1037 71

View Full Product Details