Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTNL9
Entrez Gene ID
100125313
Gene Name
butyrophilin like 9
Gene Aliases
-
Chromosome Location
Chr.7: 40085164 - 40109849 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103345.1 NP_001096815.1 7 - 8 1085 65
GenBank mRNA BC149479.1 - 7 - 8 1085 65
BT029803.1 - 7 - 8 1106 65

View Full Product Details