Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THRAP3
Entrez Gene ID
505228
Gene Name
thyroid hormone receptor associated protein 3
Gene Aliases
-
Chromosome Location
Chr.3: 109552463 - 109625055 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205565.1 NP_001192494.1 11 - 12 2837 76

View Full Product Details