Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UHMK1
Entrez Gene ID
538770
Gene Name
U2AF homology motif kinase 1
Gene Aliases
-
Chromosome Location
Chr.3: 6980639 - 7000125 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205585.1 NP_001192514.1 3 - 4 747 129

View Full Product Details