Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP3K19
Entrez Gene ID
538897
Gene Name
mitogen-activated protein kinase kinase kinase 19
Gene Aliases
YSK4
Chromosome Location
Chr.2: 62253314 - 62314262 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205597.1 NP_001192526.1 6 - 7 617 104

View Full Product Details