Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LINS1
Entrez Gene ID
539267
Gene Name
lines homolog 1
Gene Aliases
LINS
Chromosome Location
Chr.21: 6023688 - 6058996 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205649.1 NP_001192578.1 4 - 5 1223 90

View Full Product Details