Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD101
Entrez Gene ID
509609
Gene Name
CD101 molecule
Gene Aliases
IGSF2
Chromosome Location
Chr.3: 26201162 - 26239444 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205659.1 NP_001192588.1 7 - 8 2432 78

View Full Product Details