Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RUNDC1
Entrez Gene ID
539525
Gene Name
RUN domain containing 1
Gene Aliases
-
Chromosome Location
Chr.19: 43011971 - 43020122 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205681.3 NP_001192610.1 4 - 5 978 62

View Full Product Details