Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNAJC9
Entrez Gene ID
510898
Gene Name
DnaJ heat shock protein family (Hsp40) member C9
Gene Aliases
-
Chromosome Location
Chr.28: 29288865 - 29293350 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205686.1 NP_001192615.1 2 - 3 436 94

View Full Product Details