Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCTD8
Entrez Gene ID
539881
Gene Name
potassium channel tetramerization domain containing 8
Gene Aliases
-
Chromosome Location
Chr.6: 62834653 - 63097599 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205725.1 NP_001192654.1 1 - 2 950 72

View Full Product Details