Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELP6
Entrez Gene ID
512966
Gene Name
elongator acetyltransferase complex subunit 6
Gene Aliases
C22H3orf75, TMEM103
Chromosome Location
Chr.22: 52133185 - 52149780 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205740.1 NP_001192669.1 1 - 2 32 68

View Full Product Details